Methyl purple 0,1% w/v aqueous solution

Supplier: Thermo Fisher Scientific

Synonyms: Fleisher's Indicator Solution

42753.AP 42753.K2
42753.APEA 41.9 GBP
42753.AP 42753.K2
Methyl purple 0,1% w/v aqueous solution
Methyl purple

Physical Form: Liquid MDL No.: MFCD00070843

Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
MW: 1391.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070843
CAS Number: 1340-02-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR