STYRENE 2,5% (w/w) dispersion in water, latex microsphere 0.05 micron

Supplier: Thermo Fisher Scientific

42711.AB
42711.ABEA 424 GBP
42711.AB
STYRENE 2,5% (w/w) dispersion in water, latex microsphere 0.05 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Storage Temperature: 2°C to 8°C Physical Form: Dispersion Appearance: White Odour: Odourless MDL No.: MFCD00084450


Applications: Protein binding; size markers

Formula: C₈H₈
MW: 125000 - 250000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00084450
CAS Number: 9003-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR