Supelco® Organic Reference Standard, Benzyl Butyl Phthalate

Supplier: Merck

42438-50MG
42438-50MGEA 90.7 GBP
42438-50MG
Supelco® Organic Reference Standard, Benzyl Butyl Phthalate
Standards Organic Standards

  • GHS signal: GHS08, GHS09


Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=622bbf9f4a5fb5535365f378686e9228be7bc1e6&vid=622bbf9f4a5fb5535365f378686e9228be7bc1e6&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ300
Density 1,1 g/ml at 25 °C (lit.)
CAS No. 85-68-7
MDL No. MFCD00009440
Formula 2-[CH₃(CH₂)₃O₂C]C₆H₄CO₂CH₂C₆H₅
Molecular weight 312,36
Hazard class description H360FD - H410
Flash point 113,0 °C - closed cup
Shipping conditions STD
Storage conditions 15...25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR