Tetrahydrofuran-[D₈] Isotope_(100% D)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8

42348.KV
42348.KVEA 800 GBP
42348.KV
Tetrahydrofuran-[D₈] Isotope_(100% D)
Tetrahydrofuran-[D8]

Purity: 100% CAS No.: 1693-74-9 Molecular Formula: C4D8O Formula Weight: 80.16 Flash Point: -17°(-1°F) Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule Density: 0.985 Appearance: Colourless Melting Point: -106° Boiling Point: 66° UN No.: 2056 MDL No.: MFCD00044238

Formula: C₄D₈O
MW: 80.04 g/mol
Boiling Pt: 66 °C
Melting Pt: –106 °C
Density: 0.985
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR