2,2,4-Trimethylpentane 99,5, Environmental Grade for environmental analysis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

41904.K2
41904.K2EA 51.2 GBP
41904.K2
2,2,4-Trimethylpentane 99,5, Environmental Grade for environmental analysis
2,2,4-Trimethylpentane

Purity: 99.5+% CAS No.: 540-84-1 Molecular Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 Flash Point: -7°(19°F) Physical Form: Liquid Density: 0.692 Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943


Suitable for pesticide residue analysis

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 98…99 °C
Melting Pt: –107 °C
Density: 0.692
Flash Pt: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 14,5193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR