Platinum ≥99.9% (metals basis), gauze woven from wire 45 mesh, Ø 0.198 mm (0.0078 in)

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

41814.FF 41814.FI 41814.GH
41814.FFEA 389 GBP
41814.FF 41814.FI 41814.GH
Platinum ≥99.9% (metals basis), gauze woven from wire 45 mesh, Ø 0.198 mm (0.0078 in)
Platinum

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈1.61g/25x25mm, Wire Cloth Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR