Ethyl acetate 99,5, Environmental Grade for environmental analysis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Acetic acid ethyl ester

40977.K2 40977.M1
40977.K2EA 48.1 GBP
40977.K2 40977.M1
Ethyl acetate 99,5, Environmental Grade for environmental analysis
Ethyl acetate

Purity: 99.5+% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.902 Refractive Index: 1.3720 Appearance: Colourless Odour: Sweet Melting Point: -84° Boiling Point: 77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171


Pesticide residue, environmental, and GC analysis

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C
Melting Pt: –84 °C
Density: 0.902
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 14,3757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR