N-Ethylmaleimide (NEM) 99+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

40526.03 40526.06 40526.14
40526.03EA 40.3 GBP
40526.03 40526.06 40526.14
N-Ethylmaleimide (NEM) 99+%
N-Ethylmaleimide

Purity: 99+% CAS No.: 128-53-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Flash Point: 73°(163°F) Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: White to pale cream Odour: Pungent Melting Point: 43-46° Boiling Point: 209-211° Beilstein Reference: 112448 Merck Reference: 14,3822 UN No.: 2928 MDL No.: MFCD00005509

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 209…211 °C
Melting Pt: 43…46 °C
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005509
CAS Number: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1 II
Merck Index: 13,03855

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR