Acetone

Supplier: Honeywell Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

40289-2.5L
40289-2.5LEA 174 GBP
40289-2.5L
Acetone
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR