Anti-Glucose Transporter-2 Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
400061-50
400061-50EA 737 GBP
400061-50
Anti-Glucose Transporter-2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Glucose Transporter-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR