Copper, gauze woven from wire 50 mesh, Ø 0.23 mm (0.009 in)

Supplier: Thermo Fisher Scientific

39713.WP 39713.WQ
39713.WPEA 59.5 GBP
39713.WP 39713.WQ
Copper, gauze woven from wire 50 mesh, Ø 0.23 mm (0.009 in)
Copper

CAS No.: 7440-50-8 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR