Acetylacetonatobis(ethylene)rhodium(I) (≥39,9% Rh)

Supplier: Thermo Fisher Scientific

39288.03 39288.MD
39288.03EA 903 GBP
39288.03 39288.MD
Acetylacetonatobis(ethylene)rhodium(I) (≥39,9% Rh)
Acetylacetonatobis(ethylene)rhodium(I)

CAS No.: 12082-47-2 Molecular Formula: C9H15O2Rh Formula Weight: 258.13 Physical Form: Crystalline Sensitivity: Air Sensitive Solubility: Soluble in dichloromethane, chloroform MDL No.: MFCD00015354

Formula: C₉H₁₆O₂Rh
MW: 259.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015354
CAS Number: 12082-47-2
EINECS: 235-147-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR