Mini Tubes, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F378570000
391-9030EA 857 GBP
391-9030
Mini Tubes, SP Bel-Art
Tubes Microtubes
Made of virgin, medical grade polypropylene.
  • Autoclavable
  • Frozen to −20 °C
  • Very good chemical resistance
Mini-Tubes offer a convenient and economical means of performing laboratory procedures using multi-channel pipettors of four, eight or twelve channels. Features an attachment bridge between tubes, forming a Mini-Tube strip; can be easily separated into single tubes by cutting apart.