Contact plates
Supplier: Greiner Bio-One
629161_600 629180_600
391-3260EA 274 GBP
391-3260 391-0258
Contact plates
Culture Dishes Petri Dishes
PS, crystal clear. Contact dishes are used in hygiene monitoring including detection of microorganisms and testing the effectiveness of cleaning and disinfection on flat surfaces by means of contact cultures. Particularly in the pharmaceutical and food industries, analysis of germ count and germ species on surfaces is of major importance to detect and identify possible sources of contamination.
  • Sterilised by gamma irradiation
  • Vented and non-vented options
  • Graduation and division into 10 mm squares to enable quick and easy analyses
Order Now
SPECIFICATIONS
Environmentally Preferable