Xylene (mixture of isomers), extra_pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dimethylbenzene

390790010 390790025
390790010.EA 92.4 GBP
390790010. 390790025.
Xylene (mixture of isomers), extra_pure
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 136…140 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.865 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥98.0 % (mixed isomers)
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR