Bromine 1 mol/l acetic acid, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

388401000 388408000
388401000.EA 179 GBP
388401000. 388408000.
Bromine 1 mol/l acetic acid, AcroSeal®
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7
Melting Pt: -7.2
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00010896
CAS Number: 7726-95-6
EINECS: 231-778-1
UN: 2922
ADR: 8 I
Merck Index: 13,01378

Order Now

Specification Test Results

Appearance Clear red Solution
Strength 0.90 to 1.10 M

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR