Sulphuric acid 93 - 98% 93-98, Environmental Grade for environmental analysis

Supplier: Thermo Fisher Scientific
Danger

38751.AP 38751.K4
38751.APEA 129 GBP
38751.AP 38751.K4
Sulphuric acid 93 - 98% 93-98, Environmental Grade for environmental analysis
Sulphuric acid

CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid, single sub-boiling quartz distillation Density: 1.84 Appearance: Colourless to pale yellow Odour: Acrid Merck Reference: 14,8974 UN No.: 1830 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,8974

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR