Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isopropylmagnesium chloride - Lithium chloride complex, Turbo Grignard

386281000 386288000
386281000.EA 211 GBP
386281000. 386288000.
Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal®
Isopropylmagnesium chloride - LiCl complex
Formula: C₃H₇ClMg·LiCl
MW: 145.24 g/mol
Density: 0.98 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD07784514
CAS Number: 745038-86-2
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear yellow to grey to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.1 to 1.5 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR