Barium fluoride, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Fisher Scientific
Warning


Ampuled under argon

38610.09 38610.18
38610.09EA 88.6 GBP
38610.09 38610.18
Barium fluoride, ultra dry ≥99.998% (metals basis)
Barium fluoride

Purity: 99.998% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: Powder, Ampouled under argon Density: 4.83 Appearance: White Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450

Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260 °C
Melting Pt: 1353 °C
Density: 4.83
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,974

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR