Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, reduced

Supplier: Thermo Fisher Scientific

38306.06 38306.14 38306.22
38306.06EA 0 GBP
38306.06 38306.14 38306.22
Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, reduced
Palladium

CAS No.: 7440-05-3 Physical Form: 900-1100m2/g MDL No.: MFCD03457879

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR