Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 aqueous solution 97 Reag. Ph. Eur. Volumetric, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

38187-1L
38187-1LEA 163 GBP
38187-1L
Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 aqueous solution 97 Reag. Ph. Eur. Volumetric, Fluka™
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Density: 1.1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
UN: 2796
ADR: 8 II
Merck Index: 13,00509

Order Now

Specification Test Results

Appearance complying
Assay 0.095 - 0.105 mol/l
Density (D 20/4) 1.083 - 1.098


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR