Diethylaluminium iodide 0.9 M toluene, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

381161000
381161000.EA 124 GBP
381161000.
Diethylaluminium iodide 0.9 M toluene, AcroSeal®
Diethylaluminium iodide
Formula: C₄H₁₀AlI
MW: 212.01
Storage Temperature: Ambient
MDL Number: MFCD00001025
CAS Number: 2040-00-8
EINECS: 218-032-0
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 19.5 to 20.5 wt % (on Al basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR