Diethylzinc 1.5 mol/l toluene, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

377311000 377318000
377311000.EA 263 GBP
377311000. 377318000.
Diethylzinc 1.5 mol/l toluene, AcroSeal®
Diethylzinc
Formula: C₄H₁₀Zn
MW: 123.51 g/mol
Density: 0.913 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Complexometric 1.4 to 1.6 M

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR