2,6-Xylenol, Supelco®

Supplier: Merck
Danger

Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene

MQ100

36715-1G
36715-1GEA 34.3 GBP
36715-1G
2,6-Xylenol, Supelco®
2,6-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 203
Melting Pt: 41…42 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1 II
Merck Index: 12,10215
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=452a9aa69a7b44bf181ca5507a1dd60fe00124c8&vid=d0d93280665675d0d1a7adbcbfb96a0e6cd1a3bc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR