Supelco® PESTANAL® Organic Reference Standard, Endosulfan Lactone

Supplier: Merck

36675-100MG
36675-100MGEA 104 GBP
36675-100MG
Supelco® PESTANAL® Organic Reference Standard, Endosulfan Lactone
Standards Organic Standards

  • GHS signal: GHS06


Caution: P301 + P310

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ac37d8cd78ab93d31a4d4a2bd1591f339c2ad68&vid=5edf056e02e9e91de22bf5efc852f6737e2d83ca&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 3868-61-9
MDL No. MFCD00168050
Formula  C₉H₄Cl₆O₂
Molecular weight 356,84
Hazard class description H301
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR