Citric acid, anhydrous 99,5 ACS

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

36664.22 36664.36 36664.A3
36664.22EA 32.4 GBP
36664.22 36664.36 36664.A3
Citric acid, anhydrous 99,5 ACS
Citric acid

Purity: 99.5+% CAS No.: 77-92-9 Molecular Formula: C6H8O7 Formula Weight: 192.13 Physical Form: Crystalline Density: 1.665 pH: 1,6 Melting Point: 153-154° Beilstein Reference: 782061 Merck Reference: 14,2326 MDL No.: MFCD00011669


Vitamin C

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153…154 °C
Density: 1.665
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 14,2326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR