2,2,4-Trimethylpentane, dry_extra over Molecular Sieve 99.5%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

364671000 364670010
364671000.EA 46.6 GBP
364671000. 364670010.
2,2,4-Trimethylpentane, dry_extra over Molecular Sieve 99.5%, AcroSeal®
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: –107 °C
Density: 0.69 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR