tri-Sodium citrate dihydrate 99,0 ACS

Supplier: Thermo Fisher Scientific

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

36439.36 36439.A3
36439.36EA 50 GBP
36439.36 36439.A3
tri-Sodium citrate dihydrate 99,0 ACS
tri-Sodium citrate dihydrate

Purity: 99.0% min CAS No.: 6132-04-3 Molecular Formula: Na3C6H5O7.2H2O Formula Weight: 294.10 (258.07anhy) Physical Form: Crystalline Appearance: Colourless Odour: Odourless Melting Point: 150° -2H2O Solubility: Soluble in water. Insoluble in alcohol Beilstein Reference: 6104939 Merck Reference: 14,8602 MDL No.: MFCD00150031


Sequestering agent, buffer, photography

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C (-2H₂O)
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 14,8602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR