Paramat Extra, Solidification point: 55 - 58 °C, in pastille form, Gurr™, with DMSO

Supplier: VWR Chemicals

361336EEA 218 GBP
361336E 361337F
Paramat Extra, Solidification point: 55 - 58 °C, in pastille form, Gurr™, with DMSO
Paraffin wax blended with synthetic polymers

Paramat Extra is a finely filtered blend of specially selected high purity paraffin wax with an addition of polymer to give improved embedding, tissue support and reliable sectioning. It contains a small amount (<1%) of DMSO which promotes improved tissue penetration with a more homogeneous matrix, especially useful for larger specimens. Residual moisture in the tissue and traces of intermedium are dispelled. The pastille shape makes for easy handling.


  • Paraffin wax blended with synthetic polymers
  • Solidification point 55 - 58 ° C
  • Paramat extra (with DMSO) has a preservation effect


Certifications: All batches are tested in an independent histology laboratory on a range of tissue specimens to ensure problem-free processing.

CAS Number: 8002-74-2

Order Now

Specification Test Results

Appearance (molten after 7 days; 65°C) Clear
Cutting/ribboning Passes test
Congealing temperature 55 - 58 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR