1,4-Dioxane 99,0 ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

360481-500ML 360481-2.5L
360481-500MLEA 84.5 GBP
360481-500ML 360481-2.5L
1,4-Dioxane 99,0 ACS, Riedel-de Haën™
1,4-Dioxane

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 102551
RTECS Number: JG8225000
UNSPSC Code: 12191602

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 100…102 °C (1.013 hPa)
Melting Pt: 12 °C
Density: 1.031…1.034 g/cm³ (20 °C)
Flash Pt: 22.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
IR spectrum complying
Assay (GC) Min. 99.0 %
Congealing - Freezing Point Min. 11.0 °C
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Titrable Acid Max. 0.0016 meq/g
Carbonyl Compounds (as HCHO) Max. 0.01 %
Peroxides (as H₂O₂) Max. 0.005 %
APHA Max. 20
Explosion Limit 22 %
Refractive Index n20/D 1.422 (lit.)
Autoignition Temperature 356 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR