Silica gel, Pore size: 60 Å 40-60 µm, ultrapure Chromatography_column

Supplier: Thermo Fisher Scientific

360050010 360050050 360050300
360050010.EA 258 GBP
360050010. 360050050. 360050300.
Silica gel, Pore size: 60 Å 40-60 µm, ultrapure Chromatography_column
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance White Powder
Particle size ≤10 % (Oversize)
Specific surface area 460 to 510 m²/g
Pore volume 0.75 to 0.80 ml/g
pH 6.5 to 7.0 (10 % suspension in water)
Volatile matter ≤6.0 % (160°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR