Zinc chloride, dry_ultra 99,999 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

ampuled under argon

35726.06 35726.14 35726.22
35726.06EA 78.3 GBP
35726.06 35726.14 35726.22
Zinc chloride, dry_ultra 99,999 Metals basis
Zinc chloride

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: Powder, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR