Gallium tribromide, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Gallium (III) bromide

Ampuled under argon

35697.03 35697.06 35697.14
35697.03EA 119 GBP
35697.03 35697.06 35697.14
Gallium tribromide, ultra dry ≥99.998% (metals basis)
Gallium tribromide

Purity: 99.998% CAS No.: 13450-88-9 Molecular Formula: GaBr3 Formula Weight: 309.45 Physical Form: Crystalline, Ampouled under argon Density: 3.69 Appearance: White Odour: Odourless Melting Point: 122° Boiling Point: 279° UN No.: 3260 MDL No.: MFCD00016100

Formula: Br₃Ga
MW: 309.44 g/mol
Boiling Pt: 279 °C
Melting Pt: 122 °C
Density: 3.69
Storage Temperature: Ambient
MDL Number: MFCD00016100
CAS Number: 13450-88-9
EINECS: 236-609-5
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR