Sulphuric acid 5% v/v aqueous solution

Supplier: Thermo Fisher Scientific
Danger

35606.K2
35606.K2EA 26.5 GBP
35606.K2
Sulphuric acid 5% v/v aqueous solution
Sulphuric acid

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid UN No.: 2796 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR