Potassium dichromate solution_standardised 0.025 N aqueous solution

Supplier: Thermo Fisher Scientific
Danger

35589.AP 35589.K2
35589.APEA 23.4 GBP
35589.AP 35589.K2
Potassium dichromate solution_standardised 0.025 N aqueous solution
Potassium dichromate

CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid MDL No.: MFCD00011367

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR