Nitric acid 1 M HNO3 (1N) Volumetric, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Nitric acid solution

35315-1L
35315-1LEA 106 GBP
35315-1L
Nitric acid 1 M HNO3 (1N) Volumetric, Fluka™
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 100 °C
Melting Pt: -42
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR