Water 99, CHROMASOLV™ Pest_Residual, Riedel-de Haën™

Supplier: Honeywell Chemicals

Synonyms: Hydrogen oxide

34478-1L
34478-1LEA 60.4 GBP
34478-1L
Water 99, CHROMASOLV™ Pest_Residual, Riedel-de Haën™
Water

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Non-volatile matter Max. 0.0005 %
Suitability for residue analysis Max. 5 ng/l
Determination of P/N ≤5 ng/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR