Zinc sulphate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Warning

34448-1L
34448-1LEA 120 GBP
34448-1L
Zinc sulphate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
MW: 287.56 g/mol
Boiling Pt: 100 °C (solvent)
Melting Pt: <0 °C
Density: 1.005 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7446-20-0
EINECS: 231-793-3

Order Now

Specification Test Results

Factor 0.998 - 1.002

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR