, Hydranal™ Drying_agent, KF titration_air and gas, Ø ~2 mm, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

34241-250G
34241-250GEA 61.4 GBP
34241-250G
, Hydranal™ Drying_agent, KF titration_air and gas, Ø ~2 mm, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Density: 2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 1318-02-1

Order Now

Specification Test Results

Suitability (for KF-titration) test passed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR