Dimetridazole-[1-methyl-D3], Supelco®

Supplier: Merck


MQ100

34196-10MG
34196-10MGEA 208 GBP
34196-10MG
Dimetridazole-[1-methyl-D3], Supelco®
Dimetridazole-[1-methyl-D3]
Formula: C₅H₄D₃N₃O₂
MDL Number: MFCD09265282
CAS Number: 64678-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3563dfc7706f80ae15a8e3526724e5fa804e796d&vid=cebe077f9221a9da2e7f4c3068f3dad425d04211&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR