2-Hydroxydimetridazole, Supelco®

Supplier: Merck

Synonyms: (1-Methyl-5-nitro-1H-imidazol-2-yl)methanol

MQ100

34003-10MG-R
34003-10MG-REA 496 GBP
34003-10MG-R
2-Hydroxydimetridazole, Supelco®
2-Hydroxydimetridazole
Formula: C₅H₇N₃O₃
MW: 157.1274 g/mol
MDL Number: MFCD00159675
CAS Number: 936-05-0
EINECS: 213-312-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d96a96a0645987f914152c0bdbf2edc05ff6cf24&vid=4355a205da4539db7e934321931e669d9c80cb2f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR