Supelco® PESTANAL® Organic Reference Standard, α-HCH

Supplier: Merck

33856-50MG
33856-50MGEA 71.5 GBP
33856-50MG
Supelco® PESTANAL® Organic Reference Standard, α-HCH
Standards Organic Standards

  • GHS signal: GHS08, GHS06, GHS09


Caution: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aaa0a71de2574cd3c2c6c407b00b3bd72d01add&vid=d09a6191055d37132b97a63bcd16cbe2dfe4f11d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) α-1,2,3,4,5,6-Hexachlorocyclohexane; α-BHC
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 319-84-6
MDL No. MFCD00135946
Formula C₆H₆Cl₆
Molecular weight 290,83
Melting point 156...161 °C (lit.)
Hazard class description H301 - H312 - H351 - H410
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR