Orthoboric acid 99,5, Gen-Apex Molecular_Biology

Supplier: VWR Chemicals
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

33601.261EA 66.3 GBP
33601.261
Orthoboric acid 99,5, Gen-Apex Molecular_Biology
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Colouration (0,1 mol/l; water) Max. 10 APHA
Heavy metals (as Pb) Max. 5 ppm
Transmittance (230 nm) (0,1 mol/l) Min. 99 %
Transmittance (260 nm) (0,1 mol/l) Min. 95 %
Transmittance (280 nm) (0,1 mol/l) Min. 97 %
Transmittance (320 nm) (0.1 mol/l) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR