2-Propanol 99,8 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

33539-155KG 33539-1L-GL 33539-1L 33539-2.5L 33539-25L 33539-4X2.5L-G 33539-4X2.5L 33539-5L 33539-6X1L 33539-2.5L-GL 33539-200L 33539-18L
33539-155KGEA 7400 GBP
33539-155KG 33539-1L-GL 33539-1L 33539-2.5L 33539-25L 33539-4X2.5L-GL 33539-4X2.5L 33539-5L 33539-6X1L 33539-2.5L-GL 33539-200L HONC33539-18L
2-Propanol 99,8 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
2-Propanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635639
RTECS Number: NT8050000
UNSPSC Code: 12191601

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 77…83 °C (1.013 hPa)
Melting Pt: –90 °C
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 44 mmHg ( 25 °C)
Assay (GC) Min. 99.8 %
Boiling range 81 - 83 °C
Density (D 20/4) 0.784 - 0.786
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₂H₅COOH) Max. 0.00074 %
Free alkali (as NH₃) Max. 0.00017 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.0001 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
APHA Max. 10
Carbonyl compounds (as CO) Max. 0.005 %
Carbonyl compounds (ACS) complying
Ethanol (GC) Max. 0.01 %
Methanol (GC) Max. 0.1 %
KMnO₄ reducing matter (as O) Max. 0.0005 %
Solubility in water complying
Peroxides (as H₂O₂) Max. 0.00034 %
Reaction against H₂SO₄ complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR