Acetic acid glacial_AcOH 99

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

33252.AK 33252.K2 33252.M4 33252.M6
33252.AKEA 36.8 GBP
33252.AK 33252.K2 33252.M4 33252.M6
Acetic acid glacial_AcOH 99
Acetic acid

Purity: 99+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118.1 °C
Melting Pt: 16.6 °C
Density: 1.049
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
Merck Index: 14,55
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR