Combustion trays, ceramic, AL23
Supplier: Thermo Fisher Scientific
33183.KF 33184.KF 33185.KF 33186.KF
33183.KFEA 72.1 GBP
33183.KF 33184.KF 33185.KF 33186.KF
Combustion trays, ceramic, AL23
Boats Combustion Boats
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.