Supelco® PESTANAL® Organic Reference Standard, Orthosulfamuron

Supplier: Merck

32427-50MG
32427-50MGEA 378 GBP
32427-50MG
Supelco® PESTANAL® Organic Reference Standard, Orthosulfamuron
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ae4425cc0c326e521a3110e5112da7e2b424aeb&vid=e86644f5405f4e59e1556e0d544208fb3759dde5&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) 1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[2-(dimethylcarbamoyl)phenylsulfamoyl]urea
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 213464-77-8
MDL No. MFCD09027340
Formula C₁₆H₂₀N₆O₆S
Molecular weight 424,43
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR