di-Ammonium oxalate monohydrate 99,5-101,0, Puriss. p.a. ISO, ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Chemicals
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

32304-1KG 32304-250G 32304-500G
32304-1KGEA 117 GBP
32304-1KG 32304-250G 32304-500G
di-Ammonium oxalate monohydrate 99,5-101,0, Puriss. p.a. ISO, ACS, Reag. Ph. Eur., Fluka™
di-Ammonium oxalate monohydrate
Formula: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
Boiling Pt: 70
Melting Pt: 133 °C
Density: 1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
Merck Index: 13,00541

Order Now

Specification Test Results

Assay 99.5 - 101.0 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.02 %
pH (2.5 %, 20 °C) 6-7
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0002 %
Potassium (K) Max. 0.001 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.001 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Nitrate (NO₃) Max. 0.0005 %
Sulfate (SO₄) Max. 0.002 %
Solution 10 g to 200 mL (H₂O) complying

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR