Sodium hydroxide concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

Dose for preparation of 1 litre of solution.

32067.600EA 28.5 GBP
32067.600
Sodium hydroxide concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: >105 °C (1013 hPa)
Melting Pt: 323
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Titer (20°C) 0.0995 to 0.1005 mol/l
Conforms to BDH 18043 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR