Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetric solution, for titration in non-aqueous media, Fluka™

Supplier: Honeywell Chemicals
Danger

32046-6X1EA 32046-1EA
32046-6X1EAEA 439 GBP
32046-6X1EA 32046-1EA
Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetric solution, for titration in non-aqueous media, Fluka™
Perchloric acid

Concentrate for 1L standard solution

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: –18 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR